4-Bromo-5-fluoro-2,3-dihydro-1H-inden-1-one ≥97%

Lieferant: Apollo Scientific

PC540022-250MG
APOSPC540022-250MGEA 115 EUR
APOSPC540022-250MG
4-Bromo-5-fluoro-2,3-dihydro-1H-inden-1-one ≥97%
4-Bromo-5-fluoro-2,3-dihydro-1H-inden-1-one
Formel: C₉H₆BrFO
Molecular Weight: 229,046 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18208170
CAS-Nummer: 935681-01-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR