3-Bromo-5-fluoro-2-methylbenzoic acid > 97%

Lieferant: Apollo Scientific

PC540023-250MG
APOSPC540023-250MGEA 88.6 EUR
APOSPC540023-250MG
3-Bromo-5-fluoro-2-methylbenzoic acid > 97%
3-Bromo-5-fluoro-2-methylbenzoic acid
Formel: C₈H₆BrFO₂
Molecular Weight: 233,034 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19440362
CAS-Nummer: 1187318-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR