4-Bromo-2-fluoro-5-methoxybenzaldehyde ≥97%

Lieferant: Apollo Scientific

PC540024-250MG
APOSPC540024-250MGEA 79.9 EUR
APOSPC540024-250MG
4-Bromo-2-fluoro-5-methoxybenzaldehyde ≥97%
4-Bromo-2-fluoro-5-methoxybenzaldehyde
Formel: C₈H₆BrFO₂
Molecular Weight: 233,034 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD17012743
CAS-Nummer: 749931-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR