Methyl 2-chloro-6-(trifluoromethyl)isonicotinate ≥98%

Lieferant: Apollo Scientific

PC540026-250MG
APOSPC540026-250MGEA 125 EUR
APOSPC540026-250MG
Methyl 2-chloro-6-(trifluoromethyl)isonicotinate ≥98%
Methyl 2-chloro-6-(trifluoromethyl)isonicotinate
Formel: C₈H₅ClF₃NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD16611605
CAS-Nummer: 1227594-40-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR