7-Bromo-6-fluoro-2,3-dihydro-4H-1-benzopyran-4-one ≥95%

Lieferant: Apollo Scientific

Synonyme: 7-Bromo-6-fluorochroman-4-one

PC540027-250MG
APOSPC540027-250MGEA 248 EUR
APOSPC540027-250MG
7-Bromo-6-fluoro-2,3-dihydro-4H-1-benzopyran-4-one ≥95%
7-Bromo-6-fluoro-2,3-dihydro-4H-1-benzopyran-4-one
Formel: C₉H₆BrFO₂
Molecular Weight: 245,045 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18648382
CAS-Nummer: 27407-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR