6-Bromo-8-fluorochroman-4-one ≥97%

Lieferant: Apollo Scientific

PC540028-250MG
APOSPC540028-250MGEA 89.9 EUR
APOSPC540028-250MG
6-Bromo-8-fluorochroman-4-one ≥97%
6-Bromo-8-fluorochroman-4-one
Formel: C₉H₆BrFO₂
Molecular Weight: 245,045 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11207542
CAS-Nummer: 1092348-68-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR