7-Bromo-8-fluorochroman-4-one ≥97%

Lieferant: Apollo Scientific

PC540029-250MG
APOSPC540029-250MGEA 148 EUR
APOSPC540029-250MG
7-Bromo-8-fluorochroman-4-one ≥97%
7-Bromo-8-fluorochroman-4-one
Formel: C₉H₆BrFO₂
Molecular Weight: 245,045 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11518510
CAS-Nummer: 1092350-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR