6-Chlor-2-fluor-3-iodpyridin ≥97%
Lieferant: Apollo Scientific
PC540030-250MG
APOSPC540030-250MGEA
128
EUR
APOSPC540030-250MG
6-Chlor-2-fluor-3-iodpyridin ≥97%
6-Chlor-2-fluor-3-iodpyridin
Formel:
C₅H₂ClFIN Molecular Weight: 257,43 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12922962 CAS-Nummer: 1187732-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...