4-Aminobenzene-1-sulphonyl fluoride ≥97%
Lieferant: Apollo Scientific
PC540032-250MG
APOSPC540032-250MGEA
236
EUR
APOSPC540032-250MG
4-Aminobenzene-1-sulphonyl fluoride ≥97%
4-Aminobenzene-1-sulphonyl fluoride
Formel:
C₆H₆FNO₂S Lagertemperatur: Kühlschrank |
MDL:
MFCD00007883 CAS-Nummer: 98-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...