3,5-Dichloro-2-fluorobenzaldehyde ≥97%

Lieferant: Apollo Scientific

PC540033-250MG
APOSPC540033-250MGEA 75.4 EUR
APOSPC540033-250MG
3,5-Dichloro-2-fluorobenzaldehyde ≥97%
3,5-Dichloro-2-fluorobenzaldehyde
Formel: C₇H₃Cl₂FO
Molecular Weight: 193.003 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD26687657
CAS-Nummer: 477535-42-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR