6-Bromo-2-chloro-3-fluorophenol ≥97%

Lieferant: Apollo Scientific

PC540035-250MG
APOSPC540035-250MGEA 92.8 EUR
APOSPC540035-250MG
6-Bromo-2-chloro-3-fluorophenol ≥97%
6-Bromo-2-chloro-3-fluorophenol
Formel: C₆H₃BrClFO
Molecular Weight: 225,443 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28734041
CAS-Nummer: 1805518-71-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR