6-Amino-2-bromo-3-fluoro-benzoic acid ≥98%
Lieferant: Apollo Scientific
PC540036-250MG
APOSPC540036-250MGEA
348
EUR
APOSPC540036-250MG
6-Amino-2-bromo-3-fluoro-benzoic acid ≥98%
6-Amino-2-bromo-3-fluoro-benzoic acid
Formel:
C₇H₅BrFNO₂ Molecular Weight: 234.023 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28398047 CAS-Nummer: 1623461-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...