4-Bromo-2-(difluoromethoxy)-6-methylpyridine ≥97%

Lieferant: Apollo Scientific

PC540037-250MG
APOSPC540037-250MGEA 284 EUR
APOSPC540037-250MG
4-Bromo-2-(difluoromethoxy)-6-methylpyridine ≥97%
4-Bromo-2-(difluoromethoxy)-6-methylpyridine
Formel: C₇H₆BrF₂NO
Molecular Weight: 238,029 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21605151
CAS-Nummer: 1227184-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR