3-Fluoro-2-methoxybenzeneboronic acid pinacol ester ≥98%

Lieferant: Apollo Scientific

PC540040-250MG
APOSPC540040-250MGEA 142 EUR
APOSPC540040-250MG
3-Fluoro-2-methoxybenzeneboronic acid pinacol ester ≥98%
3-Fluoro-2-methoxybenzeneboronic acid pinacol ester
Formel: C₁₃H₁₈BFO₃
Lagertemperatur: Kühlschrank
MDL: MFCD16996297
CAS-Nummer: 1417301-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR