1-(4-Bromophenyl)-2,2,2-trifluoroethanol ≥96%

Lieferant: Apollo Scientific

PC540041-250MG
APOSPC540041-250MGEA 68.3 EUR
APOSPC540041-250MG
1-(4-Bromophenyl)-2,2,2-trifluoroethanol ≥96%
1-(4-Bromophenyl)-2,2,2-trifluoroethanol
Formel: C₈H₆BrF₃O
Molecular Weight: 255,032 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12786801
CAS-Nummer: 76911-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR