1-Bromo-3,4,5-trifluoro-2-nitrobenzene ≥97%

Lieferant: Apollo Scientific

PC540042-250MG
APOSPC540042-250MGEA 34.8 EUR
APOSPC540042-250MG
1-Bromo-3,4,5-trifluoro-2-nitrobenzene ≥97%
1-Bromo-3,4,5-trifluoro-2-nitrobenzene
Formel: C₆HBrF₃NO₂
Molecular Weight: 255,977 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21603959
CAS-Nummer: 1416373-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR