(5-Fluoro-2-oxo-1,2-dihydropyridin-4-yl)boronic acid > 98%
Lieferant: Apollo Scientific
PC540043-250MG
APOSPC540043-250MGEA
208
EUR
APOSPC540043-250MG
(5-Fluoro-2-oxo-1,2-dihydropyridin-4-yl)boronic acid > 98%
(5-Fluoro-2-oxo-1,2-dihydropyridin-4-yl)boronic acid
Formel:
C₅H₅BFNO₃ Molecular Weight: 156,908 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD16611113 CAS-Nummer: 1427305-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...