(4-(Aminomethyl)-3-fluorophenyl)boronic acid ≥97%

Lieferant: Apollo Scientific

PC540044-250MG
APOSPC540044-250MGEA 243 EUR
APOSPC540044-250MG
(4-(Aminomethyl)-3-fluorophenyl)boronic acid ≥97%
(4-(Aminomethyl)-3-fluorophenyl)boronic acid
Formel: C₇H₉BFNO₂
Molecular Weight: 168,961 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD24444629
CAS-Nummer: 1073055-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR