2-Chloro-4-ethyl-5-fluoropyrimidine ≥98%
Lieferant: Apollo Scientific
PC540046-100MG
APOSPC540046-100MGEA
391
EUR
APOSPC540046-100MG
2-Chloro-4-ethyl-5-fluoropyrimidine ≥98%
2-Chloro-4-ethyl-5-fluoropyrimidine
Formel:
C₆H₆ClFN₂ Molecular Weight: 160,577 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16659066 CAS-Nummer: 137234-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...