2-Amino-3-(difluoromethoxy)phenol ≥98%

Lieferant: Apollo Scientific

PC540048-100MG
APOSPC540048-100MGEA 627 EUR
APOSPC540048-100MG
2-Amino-3-(difluoromethoxy)phenol ≥98%
2-Amino-3-(difluoromethoxy)phenol
Formel: C₇H₇F₂NO₂
Molecular Weight: 175,133 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32639683
CAS-Nummer: 1597140-36-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR