8-Fluoro-1-naphthoic acid ≥98%

Lieferant: Apollo Scientific

PC540051-100MG
APOSPC540051-100MGEA 734 EUR
APOSPC540051-100MG
8-Fluoro-1-naphthoic acid ≥98%
8-Fluoro-1-naphthoic acid
Formel: C₁₁H₇FO₂
Molecular Weight: 190,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15527564
CAS-Nummer: 405196-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR