6-Chloro-8-fluoroquinazolin-4(3H)-one ≥98%

Lieferant: Apollo Scientific

Synonyme: 6-Chloro-8-fluoroquinazolin-4(1H)-one

PC540052-100MG
APOSPC540052-100MGEA 243 EUR
APOSPC540052-100MG
6-Chloro-8-fluoroquinazolin-4(3H)-one ≥98%
6-Chloro-8-fluoroquinazolin-4(3H)-one
Formel: C₈H₄ClFN₂O
Molecular Weight: 198,582 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 187805-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR