Ethyl 6-fluorobenzofuran-3-carboxylate ≥98%
Lieferant: Apollo Scientific
PC540054-100MG
APOSPC540054-100MGEA
412
EUR
APOSPC540054-100MG
Ethyl 6-fluorobenzofuran-3-carboxylate ≥98%
Ethyl 6-fluorobenzofuran-3-carboxylate
Formel:
C₁₁H₉FO₃ Molecular Weight: 208,186 g/mol Lagertemperatur: Raumtemperatur |
CAS-Nummer:
863250-72-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...