5-Chloro-7-(trifluoromethyl)imidazo[1,5-a]pyridine ≥98%

Lieferant: Apollo Scientific

PC540055-100MG
APOSPC540055-100MGEA 753 EUR
APOSPC540055-100MG
5-Chloro-7-(trifluoromethyl)imidazo[1,5-a]pyridine ≥98%
5-Chloro-7-(trifluoromethyl)imidazo[1,5-a]pyridine
Formel: C₈H₄ClF₃N₂
Molecular Weight: 220,579 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2064217-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR