6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine ≥98%
Lieferant: Apollo Scientific
PC540056-100MG
APOSPC540056-100MGEA
0
EUR
APOSPC540056-100MG
6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine ≥98%
6-Chloro-3-(trifluoromethyl)-1H-pyrazolo[4,3-c]pyridine
Formel:
C₇H₃ClF₃N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28677466 CAS-Nummer: 1431720-68-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...