6-Chloro-5-fluoroisoindolin-1-one hydrochloride ≥98%
Lieferant: Apollo Scientific
PC540057-100MG
APOSPC540057-100MGEA
453
EUR
APOSPC540057-100MG
6-Chloro-5-fluoroisoindolin-1-one hydrochloride ≥98%
6-Chloro-5-fluoroisoindolin-1-one hydrochloride
Formel:
C₈H₅ClFNO·ClH Molecular Weight: 222,044 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640848 CAS-Nummer: 2438637-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...