2,2-Difluoro-2-(quinolin-6-yl)acetic acid ≥98%

Lieferant: Apollo Scientific

PC540058-100MG
APOSPC540058-100MGEA 0 EUR
APOSPC540058-100MG
2,2-Difluoro-2-(quinolin-6-yl)acetic acid ≥98%
2,2-Difluoro-2-(quinolin-6-yl)acetic acid
Formel: C₁₁H₇F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19688050
CAS-Nummer: 1093341-40-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR