5-Bromo-2-fluoroisophthalonitrile ≥98%

Lieferant: Apollo Scientific

PC540059-100MG
APOSPC540059-100MGEA 521 EUR
APOSPC540059-100MG
5-Bromo-2-fluoroisophthalonitrile ≥98%
5-Bromo-2-fluoroisophthalonitrile
Formel: C₈H₂BrFN₂
Molecular Weight: 225.017 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28141754
CAS-Nummer: 1250411-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR