6-Bromo-8-fluoroisoquinoline ≥98%
Lieferant: Apollo Scientific
PC540061-100MG
APOSPC540061-100MGEA
112
EUR
APOSPC540061-100MG
6-Bromo-8-fluoroisoquinoline ≥98%
6-Bromo-8-fluoroisoquinoline
Formel:
C₉H₅BrFN Molecular Weight: 226.045 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28125257 CAS-Nummer: 1258833-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...