6-Bromo-5-fluoroisoindolin-1-one ≥98%
Lieferant: Apollo Scientific
PC540062-100MG
APOSPC540062-100MGEA
641
EUR
APOSPC540062-100MG
6-Bromo-5-fluoroisoindolin-1-one ≥98%
6-Bromo-5-fluoroisoindolin-1-one
Formel:
C₈H₅BrFNO Molecular Weight: 230,034 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23707957 CAS-Nummer: 1427454-77-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...