8-Bromo-3-fluoroquinolin-2(1H)-one ≥98%
Lieferant: Apollo Scientific
Synonyme:
8-Bromo-3-fluoroquinolin-2-ol
PC540063-100MG
APOSPC540063-100MGEA
753
EUR
APOSPC540063-100MG
8-Bromo-3-fluoroquinolin-2(1H)-one ≥98%
8-Bromo-3-fluoroquinolin-2(1H)-one
Formel:
C₉H₅BrFNO Molecular Weight: 242,045 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD30177936 CAS-Nummer: 834883-96-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...