(4-Fluorobenzo[d]thiazol-2-yl)methanol ≥98%

Lieferant: Apollo Scientific

PC540064-100MG
APOSPC540064-100MGEA 230 EUR
APOSPC540064-100MG
(4-Fluorobenzo[d]thiazol-2-yl)methanol ≥98%
(4-Fluorobenzo[d]thiazol-2-yl)methanol
Formel: C₈H₆FNOS
Molecular Weight: 183,203 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1188025-81-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR