Ethyl 5-chloro-7-fluoro-1H-indole-2-carboxylate ≥98%

Lieferant: Apollo Scientific

PC540065-100MG
APOSPC540065-100MGEA 264 EUR
APOSPC540065-100MG
Ethyl 5-chloro-7-fluoro-1H-indole-2-carboxylate ≥98%
Ethyl 5-chloro-7-fluoro-1H-indole-2-carboxylate
Formel: C₁₁H₉ClFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD22205716
CAS-Nummer: 1204501-37-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR