7-Bromo-4,6-dichloro-8-fluoroquinazoline ≥98%
Lieferant: Apollo Scientific
PC540066-100MG
APOSPC540066-100MGEA
0
EUR
APOSPC540066-100MG
7-Bromo-4,6-dichloro-8-fluoroquinazoline ≥98%
7-Bromo-4,6-dichloro-8-fluoroquinazoline
Formel:
C₈H₂BrCl₂FN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD29058769 CAS-Nummer: 1698027-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...