3-Chloro-2-fluoro-4-nitrophenol ≥98%

Lieferant: Apollo Scientific

PC540067-100MG
APOSPC540067-100MGEA 84.1 EUR
APOSPC540067-100MG
3-Chloro-2-fluoro-4-nitrophenol ≥98%
3-Chloro-2-fluoro-4-nitrophenol
Formel: C₆H₃ClFNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28752292
CAS-Nummer: 1805115-08-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR