6-Fluoro-2-morpholinonicotinic acid ≥98%
Lieferant: Apollo Scientific
PC540068-100MG
APOSPC540068-100MGEA
48
EUR
APOSPC540068-100MG
6-Fluoro-2-morpholinonicotinic acid ≥98%
6-Fluoro-2-morpholinonicotinic acid
Formel:
C₁₀H₁₁FN₂O₃ Molecular Weight: 226,204 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640841 CAS-Nummer: 2070856-05-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...