5-Bromo-2-(piperidin-1-yl)-3-(trifluoromethyl)pyridine ≥98%
Lieferant: Apollo Scientific
PC540069-100MG
APOSPC540069-100MGEA
260
EUR
APOSPC540069-100MG
5-Bromo-2-(piperidin-1-yl)-3-(trifluoromethyl)pyridine ≥98%
5-Bromo-2-(piperidin-1-yl)-3-(trifluoromethyl)pyridine
Formel:
C₁₁H₁₂BrF₃N₂ Molecular Weight: 309,126 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27665031 CAS-Nummer: 2151398-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...