Ethyl 2,3-diamino-4-fluorobenzoate ≥98%

Lieferant: Apollo Scientific

PC540070-100MG
APOSPC540070-100MGEA 4670 EUR
APOSPC540070-100MG
Ethyl 2,3-diamino-4-fluorobenzoate ≥98%
Ethyl 2,3-diamino-4-fluorobenzoate
Formel: C₉H₁₁FN₂O₂
Molecular Weight: 198,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28503762
CAS-Nummer: 1936001-93-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR