5-Fluoro-4-methoxypyrimidine ≥98%

Lieferant: Apollo Scientific

PC540071-250MG
APOSPC540071-250MGEA 116 EUR
APOSPC540071-250MG
5-Fluoro-4-methoxypyrimidine ≥98%
5-Fluoro-4-methoxypyrimidine
Formel: C₅H₅FN₂O
Molecular Weight: 128,104 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28163529
CAS-Nummer: 120258-30-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR