5-(Benzyloxy)-2-fluoropyridine ≥98%

Lieferant: Apollo Scientific

Synonyme: 5-Benzyloxy-2-fluoropyridine

PC540073-100MG
APOSPC540073-100MGEA 179 EUR
APOSPC540073-100MG
5-(Benzyloxy)-2-fluoropyridine ≥98%
5-(Benzyloxy)-2-fluoropyridine
Formel: C₁₂H₁₀FNO
Molecular Weight: 203.212 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24171051
CAS-Nummer: 1204483-95-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR