1-Ethyl-2-fluoro-4-methoxy-5-nitrobenzene ≥98%
Lieferant: Apollo Scientific
PC540075-1G
APOSPC540075-1GEA
412
EUR
APOSPC540075-1G
1-Ethyl-2-fluoro-4-methoxy-5-nitrobenzene ≥98%
1-Ethyl-2-fluoro-4-methoxy-5-nitrobenzene
Formel:
C₉H₁₀FNO₃ Molecular Weight: 199,179 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16658623 CAS-Nummer: 1089282-52-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...