trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride ≥98%

Lieferant: Apollo Scientific

Synonyme: trans-4-Fluoropyrrolidin-3-ol hydrochloride

PC540076-1G
APOSPC540076-1GEA 52.2 EUR
APOSPC540076-1G
trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride ≥98%
trans-4-Fluoro-3-hydroxypyrrolidine hydrochloride
Formel: C₄H₈FNO·HCl
Molecular Weight: 141,572 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23105899
CAS-Nummer: 2006333-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR