(1S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride ≥98%

Lieferant: Apollo Scientific

Synonyme: (S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride

PC540077-100MG
APOSPC540077-100MGEA 74.1 EUR
APOSPC540077-100MG
(1S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride ≥98%
(1S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride
Formel: C₅H₈F₃N·ClH
Molecular Weight: 175,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26569801
CAS-Nummer: 1338377-73-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR