2-Methoxy-5-(trifluoromethyl)aniline hydrochloride ≥98%
Lieferant: Apollo Scientific
PC540078-100MG
APOSPC540078-100MGEA
0
EUR
APOSPC540078-100MG
2-Methoxy-5-(trifluoromethyl)aniline hydrochloride ≥98%
2-Methoxy-5-(trifluoromethyl)aniline hydrochloride
Formel:
C₈H₈F₃NO·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD28167867 CAS-Nummer: 254435-23-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...