1-Bromo-2-fluoro-4-nitro-5-(trifluoromethyl)benzene ≥98%
Lieferant: Apollo Scientific
PC540080-1G
APOSPC540080-1GEA
266
EUR
APOSPC540080-1G
1-Bromo-2-fluoro-4-nitro-5-(trifluoromethyl)benzene ≥98%
1-Bromo-2-fluoro-4-nitro-5-(trifluoromethyl)benzene
Formel:
C₇H₂BrF₄NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12032178 CAS-Nummer: 1121586-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...