3-Chloro-2,4,5-trifluoro-6-nitrobenzoic acid ≥98%

Lieferant: Apollo Scientific

PC540081-250MG
APOSPC540081-250MGEA 232 EUR
APOSPC540081-250MG
3-Chloro-2,4,5-trifluoro-6-nitrobenzoic acid ≥98%
3-Chloro-2,4,5-trifluoro-6-nitrobenzoic acid
Formel: C₇HClF₃NO₄
Molecular Weight: 255,535 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25955546
CAS-Nummer: 111230-48-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR