5-Fluoro-4-iodopyridin-2(1H)-one ≥98%

Lieferant: Apollo Scientific

PC540083-100MG
APOSPC540083-100MGEA 58 EUR
APOSPC540083-100MG
5-Fluoro-4-iodopyridin-2(1H)-one ≥98%
5-Fluoro-4-iodopyridin-2(1H)-one
Formel: C₅H₃FINO
Molecular Weight: 238,986 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1227576-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR