4-(4,4,4-Trifluorobutoxy)benzoic acid

Lieferant: Apollo Scientific

PC540089-1G
APOSPC540089-1GEA 1130 EUR
APOSPC540089-1G
4-(4,4,4-Trifluorobutoxy)benzoic acid
4-(4,4,4-Trifluorobutoxy)benzoic acid
Formel: C₁₁H₁₁F₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD25710955
CAS-Nummer: 921623-31-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR