6-Hydroxy-1,1,1-trifluorohexane 95%
Lieferant: Apollo Scientific
PC540092-500MG
APOSPC540092-500MGEA
1220
EUR
APOSPC540092-500MG
6-Hydroxy-1,1,1-trifluorohexane 95%
6-Hydroxy-1,1,1-trifluorohexane
Formel:
C₆H₁₁F₃O Molecular Weight: 156.146 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02262193 CAS-Nummer: 65611-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...