3,5,7-Trihydroxy-8-(3-methyl-but-2-enyl)-2-(4-trifluoromethylphenyl)-chromen-4-one 98%

Lieferant: Apollo Scientific

Synonyme: 3,5,7-Trihydroxy-8-(3-methylbut-2-en-1-yl)-2-(4-(trifluoromethyl)phenyl)-4H-chromen-4-one

PC540096-100MG
APOSPC540096-100MGEA 836 EUR
APOSPC540096-100MG
3,5,7-Trihydroxy-8-(3-methyl-but-2-enyl)-2-(4-trifluoromethylphenyl)-chromen-4-one 98%
3,5,7-Trihydroxy-8-(3-methyl-but-2-enyl)-2-(4-trifluoromethylphenyl)-chromen-4-one
Formel: C₂₁H₁₇F₃O₅
Molecular Weight: 406.352 g/mol
MDL: MFCD28975329
CAS-Nummer: 1446712-19-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR